Veco Sjostrand for Serial Sections, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
R12CA-CU R12CA-NI
100495-976EA 68.52 CAD
100495-976 100495-978
Veco Sjostrand for Serial Sections, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco grids are the most rigid grids available and offers superior handling characteristics.
Measures 3.05 mm in diameter and with a 0.8 mil thickness.