Veco Single Slot Oval Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
2010-NI 2015-CU 2015-NI 0215-CU 0215-NI
100492-676EA 116.73 CAD
100492-676 100492-678 100492-680 100492-614 100492-616
Veco Single Slot Oval Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco grids offer superior handling characteristics.
  • Measures 3.05 mm in diameter
Veco grids are the most rigid grids available with a 0.8 mil thickness.