Single Oval Slot Grids and Slot Grids with Support Bar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G2010-NI G2010-CU G102-CU G153-CU G2010-AU G60610-CU G60610-MO G60610-NI G2010-CP
100501-528EA 102.92 CAD
100501-528 100495-668 102101-094 102101-114 102101-138 102101-196 102101-198 102101-200 103302-640
Single Oval Slot Grids and Slot Grids with Support Bar, Electron Microscopy Sciences
Graticules, Reticles and Grids
Gilder grids are a reliable support specimen grid for transmission electron microscopy.
  • Copper, copper-palladium, nickel, molybdenum, and gold
  • Grids with two central support bars enable thinner films
  • Each grid is individually inspected
Single Slot TEM Grids with oval holes have a thickness of are 50 microns.
Order Now
SPECIFICATIONS
Environmentally Preferable