Single Slot Grids (Aperture Grids), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
GA600-CU GA1500-CU GA75-CU GA600-NI GA800-NI GA1000-CU GA300-CU GA800-CU GA1000-NI GA1500-NI GA200-AU GA500-CU GA600-AU GA1000-AU GA1000-MO
100495-716EA 85.65 CAD
100495-716 100495-702 100495-718 100501-548 100501-550 100495-696 100495-712 100495-720 100495-698 100495-704 102101-234 102101-250 102101-260 103302-150 103302-194
Single Slot Grids (Aperture Grids), Electron Microscopy Sciences
Graticules, Reticles and Grids
Gilder aperture grids with a single slot.
  • Copper, nickel, molybdenum, and gold grids
  • Holes sizes of 50, 75, 100, 150, 200, 300, 400, 600, 800, 1000, 1500, and 2000 microns
  • 50 micron thickness
Single Slot aperture TEM grids have a thickness of are 50 microns. Each grid is individually inspected.