Veco Single Hole Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0800-CU 1000-NI 2000-CU 0600-CU 1500-CU 0800-NI 0600-NI 1000-CU 1500-NI 2000-NI
100492-638EA 100.52 CAD
100492-638 100492-648 100492-652 100492-650 100492-672 100492-640 102092-226 100492-646 100492-668 100492-666
Veco Single Hole Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics.
Veco Grids are the most rigid grids available. Measures 3.05 mm in diameter. Material: Copper (Cu); Nickel (Ni).
With a 0.8 mil thickness.