SEM Single Mount Storage Tube, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76530-10 76530-05 76530-01
100489-614EA 170.77 CAD
100489-614 100501-242 100501-916
SEM Single Mount Storage Tube, Electron Microscopy Sciences
Mailers and Shippers
A plastic tube and plug for storing SEM pin stub mounts. The pin mount 3.1 mm (1/8") in diameter is inserted securely to the plug and protected by the tube. Ideal for storage or mailing.