Seeker/Probe, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72945-01 72945-02
100493-910EA 21.85 CAD
100493-910 100493-912
Seeker/Probe, Electron Microscopy Sciences
Dissection Needles
Stainless steel with a hexagonal handle
Single end probe which comes with one fine point curved tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip.