Sedgewick Rafter (Microlite) Cell, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68050-51 68050-50 68050-52
100502-552EA 50.73 CAD
100502-552 100499-392 100499-394
Sedgewick Rafter (Microlite) Cell, Electron Microscopy Sciences
Cover Glasses
For use in water analysis, culture inspection, and any fluid where particles per unit volume need to be determined. Available in two versions.
S50 is an economically priced plastic cell. S52 is made from glass, with a chromium surface image. This is intended for professional use and when using phase contrast. In each type, a central cell is filled with fluid and a coverglass limits the volume to 1 milliliter. The grid subdivides this into micro-liters. S51 is the spare cover glass for S50 and S52. Each cell is supplied with one cover glass.