Scotch Conductive Tapes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77800 77802-22 77802 77798 77799 77801
100492-336EA 87.45 CAD
100492-336 100502-870 100492-338 102091-341 102091-343 102091-345
Scotch Conductive Tapes, Electron Microscopy Sciences
Tapes
Copper and aluminum conductive tapes with continuous long term usage up to 155 °C
  • Functional days-weeks usage up to 175 °C
  • Copper double sided available
We offer two types of foil: Aluminum foil tape (Scotch 1170) and copper foil tape (Scotch 1181) with conductive adhesive. 3” core, 1/4" (6.35 mm) width×54 ft (16.5 m) long.