Eyepiece Graticules, Scales with Crosslines, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68012-21 68013-21 68013-16 68012-19 68013-19 68012-16 67980-16 67980-19 67980-21 67980-50 68013-20.4 68013-23 68013-24.5 68013-25 68013-26 68013-27
100499-156EA 319.71 CAD
100499-156 100499-162 100499-158 100499-154 100499-160 100499-152 102096-042 102096-044 102096-046 102096-048 103305-056 103305-058 103305-060 103305-062 103305-064 103305-066
Eyepiece Graticules, Scales with Crosslines, Electron Microscopy Sciences
Graticules, Reticles and Grids
Useful when interested in measurement in different axis.
  • Used as a horizontal and a vertical scale
The microscope graticle is used as a horizontal and a vertical scale and is especially useful when measuring two axis.
NE7 : Horizontal micrometer scale 10 mm (0.393 inches) long with crosslines at 0.1 mm (0.003 inches) spacing.
NE77 : Horizontal micrometer scale 5 mm (0.196 inches) long in 100 divisions with crosslines.
NE777 : Horizontal micrometer scale12.7 mm (0.5 inches) long with divisions of 0.127 mm (0.005 inches) and crosslines.