Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63036-PS 63034-10 63034-05 63037-PP 63035-05 63035-10
100502-616CS 465.12 CAD
100502-616 100500-420 100490-276 100500-418 100500-416 100490-280 100490-278 100490-274 100500-422 100500-424
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Tubes Test Tubes
Transport vials/tubes are ideal for sample collection and storage.
  • Conical tubes with freestanding skirts
  • Tubes are autoclavable
  • Write-on area on side
Transport Vials are made from virgin polypropylene. They are available in three sizes, and come either sterile or non sterile. Graduations provide convenient and accurate references.
Transport Tubes are made from virgin polypropylene (PP) or polystyrene (PS). They also work well in Olympus automated testing equipment. Plug caps are required.
Delivery information: 5 ml and 10 ml vials are suppliedMore Product Information