Round Cover Slips (Bellco Brand), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72195-25 72195-15
100493-670EA 268.81 CAD
100493-670 100493-668 100502-208
Round Cover Slips (Bellco Brand), Electron Microscopy Sciences
Cover Glasses
Round coverslips with #1 Thickness (0.13 to 0.16 mm).
Packed 1 oz (28g) per case.