RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
24145-10 24145-25
100503-194EA 74.67 CAD
100503-194 100503-188
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
Spill Control Materials
RED-Z Solidifier will solidify, decontaminate and deodorize any water base fluid at a ratio of ¾ oz to 1 L
It contains chlorine (NaDCC) an EPA registered disinfectant - almost twice the amount recommended by the CDC. Solidifier could also be used for blood and other body fluids. RED-Z available in a pointed top bottle that fits easily in porthole containers and in a flip-top shaker top bottle for large spills.