Veco Rectangular Slotted Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
1002-CU 0502-CU 0620-NI 1002-NI
100492-654EA 156.75 CAD
100492-654 100492-634 100492-644 100492-656
Veco Rectangular Slotted Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco grids offer superior handling characteristics.
  • Measures 3.05 mm in diameter
Veco grids are the most rigid grids available with a 0.8 mil thickness.