Precision Pin Vise Handle and Diamond Stylus, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62140-05 62140-25 62140-50 62140-06 62140-07 62140-00 62140-02 62140-03 62140-04
100497-928EA 45.01 CAD
100497-928 100497-934 100497-936 100497-930 100497-932 100497-922 100497-924 100497-926 102093-944
Precision Pin Vise Handle and Diamond Stylus, Electron Microscopy Sciences
Tool Sets
This high precision pin vise handle is machined from the finest material, nonrolling gold-anodized aluminum, which makes it lightweight and versatile.
It features an 8 mm (0.31") hexagonal barrel dimension by 100 mm (4") long (overall length is 115 mm fully assembled) and it has a knurled finger-grip for positive precision control for handling and tool manipulation.
Delivery information: Comes complete with a precision drawbar for positive collets chuck clamping. The vise is supplied with a standard collet chuck opening of 2 mm (0.78") diameter and will hold a selection of our specially designed tools and collets adapter 0.8 mm (0.003") for diamond stylus chucking.