Razor Blades, Single Edge, Extra Long, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71981 71980
100491-880EA 1003.71 CAD
100491-880 100500-638
Razor Blades, Single Edge, Extra Long, Electron Microscopy Sciences
Razors and Blades
Extra long single edge razor blades are 21/4 x1/2" and 0.009" thick. Stainless steel blades have a sharp edge with better handling. Carbon steel blades are super strong and super sharp.
  • Stainless steel blades are packed 250 in a removable slide box
  • Carbon steel blades are packed 100 blades in a box