Quick Release Pipette Filler, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70957 70959 70958
100491-700EA 92.16 CAD
100491-700 100491-704 100491-698 100491-702
Quick Release Pipette Filler, Electron Microscopy Sciences
Pipettes Pipette Controllers
This pipetting device provides an easy, precise way of filling and releasing liquid.
  • Sizes are color coded
  • Fits standard or disposable pipettes and diposable serological tooled-mouthpiece pipettes as per ASTM Standard E714-80
By rotating the knurled thumb wheel the liquid is drawn up or dispensed. The soft elastic chuck has a threaded collar that secures the plastic or glass pipette to the device.