Pyrex® Plate, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71563-01 71563-06
100493-410EA 229.16 CAD
100493-410 100491-006
Pyrex® Plate, Electron Microscopy Sciences
Staining Tiles
A 9 cavity Pyrex pressed plate which offers a clear view for observation by transmitted light.
The plate measures L×W: 100×85 mm (4×3⅜"). The cavity is 6.4 mm (¼") deep with a 22 mm (⅞") opening.