Press-To-Seal Silicone Isolators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70337-74 70336-46 70339-40 70344-20 70337-76 70339-76 70345-40 70337-20 70337-44 70339-10 70336-70 70337-25 70339-05 70339-44 70336-42 70344-40 70344-70 70336-02 70336-03 70336-32 70336-33 70336-61 70336-62 70336-63 70344-10 70344-39 70344-69 70345-39 70345-68 70345-69 70346-05 70346-20 70346-44 70347-44 70348-40 70348-44 70348-70 70348-74
100490-884EA 543.93 CAD
100490-884 100490-858 100490-918 100490-928 100490-886 100490-926 100490-934 100490-870 100490-876 100490-914 100490-860 100490-872 100490-912 100502-050 100490-854 100490-930 100501-308 102096-772 102096-774 102096-780 102096-782 102096-786 102096-788 102096-790 102096-808 102096-812 102096-816 102096-824 102096-826 102096-828 102096-830 102096-834 102096-840 102096-858 102096-872 102096-876 102096-878 102096-882
Press-To-Seal Silicone Isolators, Electron Microscopy Sciences
Slides Chamber Slides
Silicone isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
  • Silicone isolators are available in silicone thickness 0.5 to 2.5 mm
  • Isolators are pre-cut into gaskets or as sheet materials with and without SecureSeal™ clean release adhesive on one or both surfaces
  • Isolators remain sealed to smooth surfaces
  • Non-adhesive isolators are autoclavable and reusable
  • Closed chambers may be formed using flexible RNase free, HybriSlip™ covers
They are available with 3 different gasket types, Silicone (S) / Silicone (S) – press-to-seal silicone gasket only, no adhesive, Silicone (S) / Adhesive (A) – press-to-seal, and Adhesive (A) / Adhesive (A) – press-to-seal adhesive on both sides. We also offer the Sheet Material that can be used to match your own design chambers.
Packaging: Isolators are available without coverslips, and packaged 25 per pack.