Precision Pin Vises, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62074-01 62074-03 62074-10 62074-02 62074-04
100497-560EA 82.33 CAD
100497-560 100497-562 100497-556 100503-054 100497-558
Precision Pin Vises, Electron Microscopy Sciences
Vises
These tool steel collets grip firmly when screw chucks are tightened.
  • The chromium-plated handle is hollow which allows for the holding of long wires
The body is stepped, with the small portion knurled allowing for rotation between the fingers. The larger portion is smooth and it easily can be held firmly in lathes and drill presses.