Portable Heat Pen: Soldering Iron, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72677-10 72677-50 72677-51
100501-456EA 252.89 CAD
100501-456 100493-826 100493-828
Portable Heat Pen: Soldering Iron, Electron Microscopy Sciences
Soldering Supplies Soldering Irons
This portable heat pen is able to heat up to 700°C and automatically recharges every time it is placed back into its recharging stand. It will last approximately 40 minutes on a full charge.
Tip length 1". It is useful for the sealing of wax onto glass knife boats and small soldering jobs around the lab. The set includes the heat pen and recharging stand.