Pop-Up Slide Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71520 71521 71522 71522-10
100491-790EA 32.63 CAD
100491-790 100496-334 102091-218 102097-588
Pop-Up Slide Holder, Electron Microscopy Sciences
Slide Holders
The Pop-Up holder holds 20 or 30 of 1 x 3" microscope slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed. Measurements: 8W x 131/2L x 11/32H".