Poly-L-Lysine Coated and Silane Treated Microscope Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63411-01 63411-02 63410-01 63410-02
100501-000BX 202.49 CAD
100501-000 100501-002 100490-312 100500-998
Poly-L-Lysine Coated and Silane Treated Microscope Slide, Electron Microscopy Sciences
Slides Adhesive Slides
Premium glass slides with a frosted end. These slides improve the adhesion of tissues to the slide. Available pre-coated with either Poly-L-Lysine or Silane (aminoalkysilane).
Order Now
SPECIFICATIONS
Environmentally Preferable