Polyethylene Vials; Poly-Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72621-10 72621-92 72621-30 72621-42 72621-22 72621-40 72621-20 72621-38 72621-12 72621-32 72621-60
100489-472EA 22.53 CAD
100489-472 100500-200 100489-476 100501-734 100500-192 100489-478 100489-474 100500-196 100500-190 100500-194 100489-480
Polyethylene Vials; Poly-Vials, Electron Microscopy Sciences
Vials
These flat bottom cylindrical vials have a a friction-fit, flip-top closure which can not be lost. Ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
  • Unbreakable
  • Withstand temps up to 90 °C
  • Wall thickness 1 mm