Polyethylene Disposable Gloves, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71095 71096 71097-L 71097-M 71097-S
102091-190EA 21.68 CAD
102091-190 102091-192 102097-322 102097-324 102097-326
Polyethylene Disposable Gloves, Electron Microscopy Sciences
Gloves
Low cost, offers good protection to hands from chemical contamination.
  • Excellent Sensitivity
  • Powderless/sulfur-free
  • Ambidextrous
  • 1¾ mil thickness
Packaging: 100 gloves per roll. Comes in a Dispenser Box.