Graduated Transfer Pipettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70962-10 70962-11S 70962-7 70962-8S 70962-8 70962-9 70962-11
101413-284EA 71.53 CAD
101413-284 101413-290 101413-292 101413-298 101413-296 101413-300 101413-288
Graduated Transfer Pipettes, Electron Microscopy Sciences
Pipettes Pasteur/Transfer Pipettes
These disposable graduated transfer pipettes are made up of polyethylene.
  • Low-affinity surface
  • Attached bulb avoids cross-contamination
  • One step biohazard disposable
These pipettes are ideal for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, etc.