Petroff-Hausser Chambers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63512-22 63512-24 63512-21 63512-25 63512-26 63512-20 63512-23
102094-784EA 2027.16 CAD
102094-784 100498-478 100498-488 100498-486 100498-484 100498-482 100498-480
Petroff-Hausser Chambers, Electron Microscopy Sciences
Slides Counting Chambers
New improved Neubauer ruling pattern with 1/400 sq. mm of ruling. Designed for the counting of bacteria, sperm, blood platelets, and the content of vaccines.
Cell is 0.02 mm deep with 1.5 mm thick glass slide allows the use off dark field microscopy.