Petri Dishe, Open with One Hand, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70662 70660 70664
100489-824EA 256.53 CAD
100489-824 100489-822 100491-284
Petri Dishe, Open with One Hand, Electron Microscopy Sciences
Culture Dishes Petri Dishes
A 50 x 9 mm petri dish design that is easily opened with one hand, yet closes to a tight seal by its unique double rim. Specimen samples are preserved until you are ready to analyze them. Humidity remains locked in. When loaded with a 47 mm absorbent pad, it is ideal for drying coated grids, removing water from grids that are supporting sections. Available with or without absorbent pads. The dishes are sterilized by Gamma Irradiation