Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71886 71888 71889 71885
100488-456EA 1049.48 CAD
100488-456 100488-458 100488-460 100488-454
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Slides Microscope Slides
Note: Long exposure to solvents will cause the slide to warp
Permanox™ is a strong, biologically inert material resistant to non-chlorinated hydrocarbons, acetone, and other EM reagents. Permanox™slides allow you to do cell fixation and staining on the slide; they can be cut with scissors. Permanox™ has not been shown to be autofluorescent. All of the microscope slides are 1 x 3".
Note: Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Xylene and TolueneMore Product Information