Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64155 64153 64157 64165 64161 64154 64162 64151
100503-302EA 31.96 CAD
100503-302 100503-268 100503-304 100503-314 100503-310 100503-300 100503-312 102091-094
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Bottles Wash Bottles
Permanently labeled/color-coded wash bottles.
Pack of six - Variety includes: Acetone, Methanol, Water, Iso-propyl Alcohol, Ethanol and Toluene. Toluene Wash Bottle is for short term storage only.
These safe wash bottles, which are permanently labeled and color-coded, eliminate the risk of dispensing the wrong liquid by mistake. All of these wide-mouth (easy transfer liquid) polyethylene bottles have polyalomer caps and a fine-tip-nozzle that may be cut back for heavier flow. Capacity is 16 oz. (500 ml), low density polyethylene.