Perfect Loop, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70945 70942 70941 70944 70940 70943 70946 70948
100491-694EA 277.42 CAD
100491-694 100489-828 100491-688 100491-692 100491-686 100491-690 100489-830 100491-696
Perfect Loop, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.