Veco Parallel Bar with Handle Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HDR300-NI HDR100-CU HDR200-CU HDR300-CU HDR100-NI HDR200-NI
100495-888EA 52.54 CAD
100495-888 100501-564 100501-566 100501-568 100495-876 100495-882
Veco Parallel Bar with Handle Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics
Veco Grids are the most rigid grids available. Measures 3.05 mm in diameter. Material: Copper (Cu); Nickel (Ni).
Plus, with a 0.8 mil thickness.