Parallel Bars Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G100PB-CU G100P-CU G100P-NI G200PB-CU G400PB-NI G400P-NI G300PB-CU G200P-NI G200PB-NI G300P-NI G100PB-NI G150P-CU G50PB-CU G75PB-CU G75P-CU
100495-626EA 72.14 CAD
100495-626 100495-630 100495-632 100501-522 100495-686 100495-690 100501-540 100502-994 100502-992 100503-004 100495-628 102101-108 102101-186 102101-214 102101-218
Parallel Bars Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt or shiny side. Each grid is individually inspected.
  • 3.05 mm diameter
  • 0.7 mil (18 µm) thickness
  • In Copper (Cu), Nickel (Ni), or Gold (Au)