Veco Parallel Bar with Divider Handle Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HDR300D-CU HDR100D-CU HDR200D-CU HDR200D-NI HDR300D-NI
100495-884EA 63.22 CAD
100495-884 100495-872 100495-878 100495-880 100495-886
Veco Parallel Bar with Divider Handle Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco grids are the most rigid grids.
  • Superior handling characteristics
  • Measures 3.05 mm in diameter with a 0.8 mil thickness
Material: Copper (Cu); Nickel (Ni).