Veco Parallel Bar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
R300-CU R300-NI R100-NI RB90-NI RB90-CU
100495-996EA 59.76 CAD
100495-996 100496-002 100495-974 100496-004 100502-352
Veco Parallel Bar, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics.
  • With a 0.8 mil thickness
  • Measures 3.05 mm in diameter
Veco Grids are the most rigid grids available. Material: Copper (Cu); Nickel (Ni).