Pap-Pen; Slide Marker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71312
71310
100502-806EA
169.93
CAD
100502-806
100491-772
Pap-Pen; Slide Marker, Electron Microscopy Sciences
Stains
Microscopy and staining reagents
New and improved staining guard pen.
New and improved staining guard pen. The Super PAP Pen outperforms the original PAP Pen. The Super PAP Pen may be used at temperatures as high as 120°F. (The old style PAP Pen worked only up to 90°F). With this high temperature feature, one can now use the Super PAP Pen to perform the peroxidase-anti-peroxidase method, ABC method, and Fluorescent antibodies method with satisfactory results.