Veco Oyster Grids, Single Slot, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
DL2010-CU DL2010-NI
100495-522EA 68.87 CAD
100495-522 100495-524
Veco Oyster Grids, Single Slot, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics.
  • With a 0.8 mil thickness
  • Measures 3.05 mm in diameter
Veco Grids are the most rigid grids available. Material: Copper (Cu); Nickel (Ni).