Veco Oyster Type Hexagonal Mesh, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
D100H-CU D75H-CU D753H-NI D200H-CU D300H-CU D300H-NI D100H-NI
100495-498EA 88.51 CAD
100495-498 100495-516 100495-512 100495-502 100495-506 100495-508 100495-500
Veco Oyster Type Hexagonal Mesh, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco grids are the most rigid grids.
  • Superior handling characteristics
  • Measures 3.05 mm in diameter with a 0.8 mil thickness
Material: Copper (Cu); Nickel (Ni)
Order Now
SPECIFICATIONS
Environmentally Preferable