Maxtaform Oval Hole Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
M2010-CR M2010-NI
100495-930EA 82.71 CAD
100495-930 100495-932
Maxtaform Oval Hole Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
High Grade Maxtaform grids with clean and smooth edges, firm support, and a large open area.
  • One surface coated with inert rhodium
  • Eliminate tarnishing
  • Reduce bar thickness
  • Give side identification
Copper grids available with 3 mm diameter, 0.75 mil thickness. Material: Copper/Rhodium (Cu/Rh=CR), Nickel (Ni), Gold (Au).