Narrow Mouth Ground Glass Stoppered Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64207-C 64204-A 64209-C 64202-C 64204-C 64206-C 64202-A
100498-646EA 755.56 CAD
100498-646 100490-410 100502-534 100490-408 100490-412 100490-416 100502-018
Narrow Mouth Ground Glass Stoppered Bottles, Electron Microscopy Sciences
Bottles Glass Bottles
New designed ground glass stoppered bottles feature a no-drip pour lid for clean pouring of contents.
The most preferred container for storage of prepared chemicals, and stock solutions. Octagon-shaped ground glass stopper is included. The borosilicate '400' glass offers a high degree of protection from leaching and the amber 500' glass  gives extra protection to light-sensitive contents.