Miller Squares Graticle NE57, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68020-19 68020-20.4
100499-208EA 343.27 CAD
100499-208 103305-392
Miller Squares Graticle NE57, Electron Microscopy Sciences
Slides Counting Chambers
The Mill Sqares microscope graticle has a ratio of large to small squares of 9:1
  • Glass sandwich
  • 7 x 7 mm grid
  • Extended size range
Designed for hematology, it can be utilized for rapid counting of any evenly spread field of particle.