MicroWell® Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70436-R1 70439-10 70437-F5 70436-ST 70438-10 70437-R1 70437-F1 70437-R5 70439-15 70436-F5 70436-L5 70436-R5 70436-F1 70436-L1
100489-136EA 170.8 CAD
100489-136 100489-144 100500-090 100500-754 100489-142 100489-140 100489-138 100500-092 100500-096 100501-728 100502-584 100500-088 100489-132 100489-134
MicroWell® Plates, Electron Microscopy Sciences
Microplates Cell Culture Plates
MicroWell Plates are used in a broad spectrum of applications, including cloning experiments, sample incubations, and in tissue culture based analytical systems. These MicroWell Plates have excellent cell attachment. All styles are designed to be stackable and are radiation sterilized.
10 µl Well Plates
They are Terasaki format plates measuring 56 x 82 mm and having 60 or 72 wells. Wells are shaped like inverted truncated cones, each with a working volume of 10 µl. The well surface is Nunclon® treated for good cell attachment. Lids incorporate a stacking feature and are tight-fitting, which reduces evaporation.
96µl Well Plates
These plates measure 86 x 128 mm and come in either flat or round bottom wells. Vented lids allow for gas exchange and have condensation rings to control evaporation and retain condensate. Lids are keyed so they cannot be accidentally reversed. Plates have alpha-numeric markings on its two edges. In addition, flat bottom plates have individual well coordinates beside each well for easy well identification when using an inverted microscope. Plates also have raised well rims to provide proper attachment of adhesive sealing film.