Microscope Slide Folder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71502-B1 71502-G1 71502-GR1 71502-W10 71502-Y1 71502-10 71502-B10 71502-GR10 71502-Y10 71502-G10 71502-W1 71502-00
100493-368EA 44.73 CAD
100493-368 100493-370 100493-372 100489-318 100493-376 100489-310 100489-312 100489-316 100489-320 100489-314 100493-374 100493-366
Microscope Slide Folder, Electron Microscopy Sciences
Slide Holders
Made from heat resistant injection molded plastic. The Slide Folder™ will hold up to twenty standard microscope slides 75 x 25 mm (3x1").
The Slide Folder™ is made of two parts: a base holding the slides horizontally offering numbered space for easy identification, and transparent doors which can either cover the slides or be swung behind the SlideFolder™ for space conservation while using the tray. The base is available in 5 different colors, offering color-coding for each job. Each slide location is identified with a number from 1 to 20. Removal of the slides is made easy by simply pressing on one end, which will lift the other end up. The two transparent hinged covers offer a full view of each stored slide, even bar codes can be read without removing of the slide. All units are stackable and will resist temperatures between –80°C and +100°C. They are not autoclavable.