Microcentrifuge Tube Racks; BestRack®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72375-05 72375-06 72375-01
100491-058EA 321.12 CAD
100491-058 100491-060 100493-764
Microcentrifuge Tube Racks; BestRack®, Electron Microscopy Sciences
Racks Tube Racks
This reversible rack holds 96 microcentrifuge tubes
  • Holds 1.5/2.0 ml tubes on one side, and 0.5 ml tubes on the other
  • Rows and columns labeled for easy sample identification
  • Frosted labeling areas on four sides
  • Withstands freezing (-90°C) and autoclaving
  • Comes in 5 popular colors as well as natural
This sturdy polypropylene rack holds 96 tubes in an 8 x 12 format. The rack provides easy access to each tube, even when 96 places are full. The transparent polypropylene lid has a snug fit and is tall enough to store most screw-cap microcentrifuge tubes.More Product Information