Metallurgical Graticles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68068-21 68069-21 68067-19 68084-19 68067-21 68068-19 68070-19 68085-19 68084-21 68069-19 68085-21 68066-21 68084-16
100499-492EA 748.65 CAD
100499-492 100499-496 100499-486 100499-504 100499-488 100499-490 100499-498 100499-510 100499-506 100499-494 100499-512 100499-484 100499-502
Metallurgical Graticles, Electron Microscopy Sciences
Graticules, Reticles and Grids
Standard pattern discs for metallurgical stereometric analysis of grain size in polished metal sections.
Order Now
SPECIFICATIONS
Environmentally Preferable