Membrane Boxes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64304-10 64304-20 64304-12 64304-39 64304-44 64304-11 64304-38 64304-17 64304-13 64304-14 64304-36 64304-41 64304-15 64304-42 64304-43 64304-45 64304-30 64304-16 64304-46 64304-50 64304-40 64304-54 64304-24 64304-25 64304-26 64304-27 64304-28 64304-29 64304-51 64304-22 64304-52 64304-57 64304-18 64304-19 64304-23 64304-58 64304-21 64304-59 64304-55 64304-56
100490-554EA 124.99 CAD
100490-554 100498-656 101412-932 101412-948 101412-958 101412-930 101412-946 101412-942 101412-934 101412-936 101412-944 101412-952 101412-938 101412-954 101412-956 101412-960 100498-658 101412-940 101412-962 100498-662 100498-660 103301-958 103301-960 103301-962 103302-034 103302-036 103302-038 103302-336 103302-338 103302-350 103302-352 103302-382 103302-386 103302-402 103302-404 103302-406 103302-420 103302-528 103302-620 103302-654
Membrane Boxes, Electron Microscopy Sciences
Mailers and Shippers
These unique membrane boxes are for the transfer, storage and shipping of many delicate items. Once your sensitive item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
  • Protects items from any movement
  • Double elastic membrane on the top and bottom offers security
  • Available in round, rectangular and square shapes