Media/Lab Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64210-C 64210-A 64214-C 64216-C 64212-C 64214-A
100500-442CS 154.95 CAD
100500-442 100500-440 100500-446 100500-448 100500-444 102094-956 102681-046
Media/Lab Bottles, Electron Microscopy Sciences
Bottles Glass Bottles
Graduated bottles have a large opening, no-drip pour lip and sloping shoulders to facilitate pouring and cleaning.
These bottles are widely used for mixing and storage of culture media, fixation and dehydrating materials. Bottles show approximate volume graduations, and have large frosted writing area. Clear bottles are Wheaton '400' borosilicate glass. Low alkali content helps to prevent changes in pH and maintain purity of contents. Comes with 'PTFE'-lined cap. Autoclavable. Amber bottles are manufactured from '500' borosilicate glass to protect light-sensitive samples.