Marker Pen, Markal Sharpie, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72120 72110 72140 72130
100500-642BX 56.3 CAD
100500-642 100491-926 100500-644 100491-928
Marker Pen, Markal Sharpie, Electron Microscopy Sciences
Pens Permanent Markers
Smear resistant, water-resistant, fast-drying no
15 fine line. Usable on glass, porcelain, metals, and plastics.