Low Temperature Hot Glue Gun, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72171-00 72171-10
100493-644EA 50.28 CAD
100493-644 100489-428
Low Temperature Hot Glue Gun, Electron Microscopy Sciences
Stains Microscopy and staining reagents
This low temperature mini glue gun is safe and allows for the easy application of the melt glue to the aluminum stub for the mounting of bulk or irregular shaped specimens for SEM.
  • Low temperature
  • Easy to use
  • 40" cord
Comes complete with a 40" cord and two glue sticks. Accepts 5/16" diameter all temperature or low temperature glue sticks. 110 volt, 10 watts.