Liquid Nitrogen-Cooled Mortar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64470 64472 64471
100496-228EA 1393.67 CAD
100496-228 101413-004 100496-230
Liquid Nitrogen-Cooled Mortar, Electron Microscopy Sciences
Mortars and Pestles Mortars
The Liquid Nitrogen-Cooled Mortar is for grinding frozen tissue samples without the hazard of liquid nitrogen droplets flying into personnel.
  • Insulated handle 4" (102 mm) is removable for saving space
  • Housing measures 61/2" (165 mm) in diameter, 41/2" high
  • Three suction cup-feet hold the housing
A stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds the liquid nitrogen. The bowl is nested in a well insulated polyethylene housing.